missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lactose Broth

Product Code. 11922642
Change view
Click to view available options
Fill Weight:
90 to 91.5 g
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight unitSize
11922642 90 to 91.5 g Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11922642 Supplier Thermo Scientific™ Supplier No. BO0596Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Presumptively identify coliform organisms in milk, water and foods with this medium.

Presumptively identify coliform organisms in milk, water and foods with Lactose Broth as specified by the American Public Health Association.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2°C to 25°C
Format 125 mL Sirop Bottle
Manufacturing Origin United Kingdom
Product Type Microbiology Media
Fill Weight 90 to 91.5 g
Packaging Type 10 Sirop Bottles with screw cap closures in a box
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.