missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactoferrin Antibody (1C6cc), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£155.00
Specifications
| Antigen | Lactoferrin |
|---|---|
| Clone | 1C6cc |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA 1:100-1:2000, CyTOF-ready |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
Description
Lactoferrin Monoclonal specifically detects Lactoferrin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA, CyTOF-ready.Specifications
| Lactoferrin | |
| Western Blot 1:100-1:2000, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA 1:100-1:2000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
| LTF | |
| IgG1 κ | |
| Protein A purified | |
| Human lactoferrin. No cross-reactivity with human transferrin. |
| 1C6cc | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Amino Acids Drugs and other small molecules | |
| P02788 | |
| 4057 | |
| Full length native protein (purified) (Human). | |
| Primary | |
| Store at 4°C. Do not freeze. | |
| 78 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title