Learn More
Lactoferrin Antibody (1C6cc), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NB120-10109-0.05mg
 View more versions of this product
Product Code. 18598911
  • £155.00 / 50µg
Description
Lactoferrin Monoclonal specifically detects Lactoferrin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lactoferrin
Monoclonal
Unconjugated
P02788
LTF
Full length native protein (purified) (Human).
Protein A purified
RUO
Primary
Human lactoferrin. No cross-reactivity with human transferrin.
Store at 4°C. Do not freeze.
IgG1 κ
Immunohistochemistry (Paraffin)
1C6cc
Western Blot 1:100-1:2000, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA 1:100-1:2000, CyTOF-ready
EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin
Mouse
78 kDa
0.05 mg
Amino Acids Drugs and other small molecules
4057
Human
Purified