missing translation for 'onlineSavingsMsg'
Learn More

Lactitol, TRC

Product Code. 30475048
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30475048 5 g 5g
30468398 1 g 1g
30466347 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30475048 Supplier Toronto Research Chemicals Supplier No. TRCL1135505G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-86-4
Molecular Formula C12 H24 O11
Molecular Weight (g/mol) 344.31
Synonym Glucitol, 4-O-β-D-galactopyranosyl-, D- (8CI), Lactitol (6CI,7CI), 4-O-β-D-Galactopyranosyl-D-glucitol, D-Lactitol, Finlac DC, Importal, Lactiobiosit, Lactit, Lactit M, Lactite, Lactitol ACM 50, Lactitol LC 0, Lactitol MC, Lactosit, Lactositol, Lacty, Lacty (saccharide), Lacty M, Lacty-Tab, Milchen, Miruhen, NSC 231323, D-Glucitol, 4-O-β-D-galactopyranosyl
IUPAC Name (2S,3R,4R,5R)-4-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol
SMILES OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)CO

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H24O11/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12/h4-21H,1-3H2/t4-,5+,6+,7+,8-,9-,10+,11+,12-/m0/s1
Formula Weight 344.1319
Percent Purity >95%
Chemical Name or Material Lactitol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.