missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactate Dehydrogenase C Antibody (9F2), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Lactate Dehydrogenase C |
|---|---|
| Clone | 9F2 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30033364
|
Novus Biologicals
NBP3-26173-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30039638
|
Novus Biologicals
NBP3-26173-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lactate Dehydrogenase C Monoclonal antibody specifically detects Lactate Dehydrogenase C in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| Lactate Dehydrogenase C | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cancer/testis antigen 32, CT32EC 1.1.1.27, EC 1.1.1, lactate dehydrogenase C, lactate dehydrogenase C4, LDH testis subunit, LDH3MGC111073, LDH-C, LDHX, LDH-X, L-lactate dehydrogenase C chain | |
| A synthesized peptide derived from Human Lactate Dehydrogenase C [UniProt P07864] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 9F2 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 3948 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title