missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactate Dehydrogenase C Antibody (9F2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26173-50ul
This item is not returnable.
View return policy
Description
Lactate Dehydrogenase C Monoclonal antibody specifically detects Lactate Dehydrogenase C in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| Lactate Dehydrogenase C | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 9F2 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Cancer/testis antigen 32, CT32EC 1.1.1.27, EC 1.1.1, lactate dehydrogenase C, lactate dehydrogenase C4, LDH testis subunit, LDH3MGC111073, LDH-C, LDHX, LDH-X, L-lactate dehydrogenase C chain | |
| A synthesized peptide derived from Human Lactate Dehydrogenase C [UniProt P07864] | |
| 50 μL | |
| Cellular Markers | |
| 3948 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction