missing translation for 'onlineSavingsMsg'
Learn More

Lactase Antibody (3G5.1/2.G6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£149.00 - £307.00

Specifications

Antigen Lactase
Clone 3G5.1/2.G6
Concentration 1 mg/mL
Dilution Immunoprecipitation 1:10 - 1:500
Applications Immunoprecipitation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18676436
View Documents
Novus Biologicals
NBP2-50512-0.025ml
0.025 mL
£149.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18601337
View Documents
Novus Biologicals
NBP2-50512
0.1 mL
£307.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lactase Monoclonal antibody specifically detects Lactase in Human, Rabbit samples. It is validated for Immunoprecipitation
Specifications

Specifications

Lactase
1 mg/mL
Immunoprecipitation
Unconjugated
Mouse
Cancer, Cellular Signaling, metabolism
PBS
3938
IgG1
Protein A purified
3G5.1/2.G6
Immunoprecipitation 1:10 - 1:500
Monoclonal
Purified
RUO
Human, Rabbit
EC 3.2.1, LAC, lactase, Lactase-glycosylceramidase, lactase-phlorizin hydrolase, LPH1, LPHlactase-phlorizin hydrolase-1
Normal jejunal epithelial brush border-enriched membranes from a non-secretor, blood group O
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.