missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactase Antibody (3G5.1/2.G6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50512-0.025ml
This item is not returnable.
View return policy
Description
Lactase Monoclonal antibody specifically detects Lactase in Human, Rabbit samples. It is validated for Immunoprecipitation
Specifications
| Lactase | |
| Monoclonal | |
| 1 mg/mL | |
| Immunoprecipitation 1:10 - 1:500 | |
| EC 3.2.1, LAC, lactase, Lactase-glycosylceramidase, lactase-phlorizin hydrolase, LPH1, LPHlactase-phlorizin hydrolase-1 | |
| Normal jejunal epithelial brush border-enriched membranes from a non-secretor, blood group O | |
| 0.025 mL | |
| Cancer, Cellular Signaling, metabolism | |
| 3938 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunoprecipitation | |
| 3G5.1/2.G6 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction