missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Holder for Glass Flasks
Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
Labor Glasses
Unit Size:
Each
Specifications
Specifications
| Type | Immersion Basket |
| Diameter (Metric) | 59 mm |
| For Use With (Equipment) | Labor Glasses |
| Height (Metric) | 60 mm |
| Material | Stainless Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction