missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ LaboPlast™ Disposable Spatula
GREENER_CHOICEBIOPHARMA

Product Code. 15336317 Shop All Bürkle Products
Click to view available options
Includes:
With Transparent Sleeve
Without Sleeve
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 15336317

Brand: Buerkle™ 53780010

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Disposable spatula features a long, ergonomically-shaped, stable handle and a sharp blade.

  • Ideal for sampling powders, granulates and pastes
  • Transparent sealing sleeve reduces contamination
  • White
  • Individually packaged
  • Production, assembly and packaging according to Cleanroom Class 7
  • With EU foodstuffs and FDA approval
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10/Pk.
Color White
Certifications/Compliance FDA
Disposable Single-use
Product Type Disposable Spatula
For Use With (Application) Ideal for Sampling Powders, Granulates, Pastes and Foodstuffs
Length (Metric) Blade 150 mm
Includes With Transparent Sleeve
Material Polystyrene
Handle Type Ergonomically shaped
Length (Metric) 268 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.