Learn More
Labconco™ Carbon Filters for Purifier™ Class I and HEPA Filtered Enclosures
For easy installation atop the Purifier Class I or HEPA Filtered Enclosures
Brand:  Labconco™ 3937400
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12822141
  • £750.00 / Each
Estimated Shipment: 19-07-2024
to see stock.
Description
Description
  • Trap nonvolatile solvent vapors/odors from samples or minute quantities of chemicals
  • Trace odor filter with organic activated carbon
Specifications
Specifications
Carbon
1 in.
5.9 Kg
18 in.
2.5 cm
45.7 cm
106.7 cm
3 ft. Purifier™ class I and HEPA filtered enclosures, XPert™ filtered balance system
13 lb.
40 to 50 in.
42 in.
40 to 50 x 42 x 1 in. (101.6 to 127 x 106.7 x 2.5 cm)
101.6 to 127 cm
Filter