Learn More
Epredia™ Lab Vision™ UltraVision™ LP Detection System: AP Polymer (Ready-To-Use)
Epredia™ Lab Vision UltraVision LP Detection System: AP Polymer is a sensitive detection system based on biotin-free, polymer technology.
£1515.00 - £2245.00
Specifications
Chromagen | None provided |
---|---|
Enzyme | Alkaline Phosphatase |
Specificity | anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L) |
Substrate | None provided |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12683997
|
Epredia™
TL-060-AL |
60mL |
£1515.00
60mL |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
11379211
|
Epredia™
TL-125-AL |
125mL |
£2245.00
125mL |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
Description
UltraVision LP is the latest technology in polymeric labeling. Polymer detection methods have been shown to provide increased sensitivity and detection simplicity. This second-generation polymer system is composed of smaller polymer subunits that minimize conflicts in binding the target protein. Decreased binding conflicts result in more consistent staining and better signal amplification. Ultimately, this gives the user higher sensitivity and antibody efficiency. With UltraVision LP, you use less antibody and obtain better signal-to-noise ratios. UltraVision LP is also biotin-free, which eliminates background staining found with traditional biotin-based detection methods.Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme: Alkaline Phosphatase
Chromogen/Substrate: None provided
Includes:
TL060AL: Ultra V Block (Mfr. No. TA-060-UB), Primary Antibody Enhancer (Mfr. No. TL-060-PB), AP Polymer (Mfr. No. TL-060-AP)
TL125AL: Ultra V Block (Mfr. No. TA-125-UB), Primary Antibody Enhancer (Mfr. No. TL-125-PB), AP Polymer (Mfr. No. TL-125-AP)
Note:
UltraVision detection system detects a specific mouse IgG or rabbit IgG antibody bound to an antigen in tissue sections. The specific antibody is located by a universal secondary antibody formulation conjugated to an enzyme-labeled polymer that recognizes mouse and rabbit immunoglobulins. The polymer complex is then visualized with an appropriate substrate/chromogen.
Specifications
None provided | |
Alkaline Phosphatase | |
anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L) | |
None provided |