missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ UltraVision™ LP Detection System: AP Polymer (Ready-To-Use)

Product Code. 11379211 Shop All Epredia Products
Change view
Click to view available options
Quantity:
60 mL
125 mL
Unit Size:
125mL
60mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379211 125 mL 125mL
12683997 60 mL 60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379211 Supplier Epredia™ Supplier No. TL125AL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Epredia™ Lab Vision UltraVision LP Detection System: AP Polymer is a sensitive detection system based on biotin-free, polymer technology.

UltraVision LP is the latest technology in polymeric labeling. Polymer detection methods have been shown to provide increased sensitivity and detection simplicity. This second-generation polymer system is composed of smaller polymer subunits that minimize conflicts in binding the target protein. Decreased binding conflicts result in more consistent staining and better signal amplification. Ultimately, this gives the user higher sensitivity and antibody efficiency. With UltraVision LP, you use less antibody and obtain better signal-to-noise ratios. UltraVision LP is also biotin-free, which eliminates background staining found with traditional biotin-based detection methods.
  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Alkaline Phosphatase

  • Chromogen/Substrate: None provided

Includes:

TL060AL: Ultra V Block (Mfr. No. TA-060-UB), Primary Antibody Enhancer (Mfr. No. TL-060-PB), AP Polymer (Mfr. No. TL-060-AP)

TL125AL: Ultra V Block (Mfr. No. TA-125-UB), Primary Antibody Enhancer (Mfr. No. TL-125-PB), AP Polymer (Mfr. No. TL-125-AP)

Note:

UltraVision detection system detects a specific mouse IgG or rabbit IgG antibody bound to an antigen in tissue sections. The specific antibody is located by a universal secondary antibody formulation conjugated to an enzyme-labeled polymer that recognizes mouse and rabbit immunoglobulins. The polymer complex is then visualized with an appropriate substrate/chromogen.

TRUSTED_SUSTAINABILITY

Specifications

Chromagen None provided
Count 625 to 1250 slides
Enzyme Alkaline Phosphatase
Specificity anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Substrate None provided
Includes Ultra V Block (Mfr. No. TA-125-UB), Primary Antibody Enhancer (Mfr. No. TL-125-PB), AP Polymer (Mfr. No. TL-125-AP)
Quantity 125 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.