Learn More
Epredia™ Lab Vision™ TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1 Mouse Monoclonal Antibody
TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1, Mouse MAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.
£126.48 - £581.40
Specifications
| Antigen | TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1 |
|---|---|
| Clone | SEN28 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12618976
|
Epredia™
MS-1105-S0 |
100 μL | Concentrated | ||||||
|
12628976
|
Epredia™
MS-1105-S1 |
500 μL | Concentrated |
£461.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12608976
|
Epredia™
MS-1105-S |
1 mL | Concentrated | ||||||
|
12688966
|
Epredia™
MS-1105-R7 |
7 mL | Prediluted | ||||||
|
12698966
|
Epredia™
MS-1105-RQ |
12 mL | Ready-to-Use | ||||||
Description
Terminal Deoxynucleotidyl Transferase (TdT) is a DNA polymerase located in the cell nucleus which catalyses the polymerization of deoxynucleotides at the 3' hydroxyl ends of oligo or polydeoxynucleotide initiators and functions without a template. TdT is considered to be a highly specific marker for the diagnosis and classification of acute lymphoblastic lymphoma/leuksemias. The determination of TdT expression is most valuable when it is different to differentiate histologically between lymphoblastic lymphoma and Burkitt's lymphoma.Host Species: Mouse
Clone: SEN28
Isotype: IgG2a
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: N-terminal
Immunogen: Recombinant protein corresponding to the amino terminal region of the TdT molecule
Molecular Weight: 58kDa
Positive Control: Thymus
Cellular Localization: Nuclear Ab-1 will be of use in the differentiation of lymphoblastic lymphoma from other lymphomas.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Recombinant protein corresponding to the amino terminal region of the TdT molecule |
| SEN28 | |
| Monoclonal | |
| Mouse | |
| DNA Repair | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.