Learn More
Epredia™ Lab Vision™ TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1 Mouse Monoclonal Antibody
TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1, Mouse MAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Brand: Epredia™ MS-1105-S1
Description
Terminal Deoxynucleotidyl Transferase (TdT) is a DNA polymerase located in the cell nucleus which catalyses the polymerization of deoxynucleotides at the 3' hydroxyl ends of oligo or polydeoxynucleotide initiators and functions without a template. TdT is considered to be a highly specific marker for the diagnosis and classification of acute lymphoblastic lymphoma/leuksemias. The determination of TdT expression is most valuable when it is different to differentiate histologically between lymphoblastic lymphoma and Burkitt's lymphoma.Host Species: Mouse
Clone: SEN28
Isotype: IgG2a
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: N-terminal
Immunogen: Recombinant protein corresponding to the amino terminal region of the TdT molecule
Molecular Weight: 58kDa
Positive Control: Thymus
Cellular Localization: Nuclear Ab-1 will be of use in the differentiation of lymphoblastic lymphoma from other lymphomas.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1 | |
| Monoclonal | |
| Unconjugated | |
| Recombinant protein corresponding to the amino terminal region of the TdT molecule | |
| IVD | |
| Primary | |
| Concentrated |
| Immunohistochemistry (Paraffin) | |
| SEN28 | |
| Mouse | |
| 500 μL | |
| DNA Repair | |
| Human | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.