missing translation for 'onlineSavingsMsg'
Learn More

Surfactant Protein B (Precursor) Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ Surfactant Protein B (Precursor) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

£130.56 - £693.60

Specifications

Antigen Surfactant Protein B (Precursor) Ab-1
Clone SPB01; same as 1-2-A5-9
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Host Species Mouse
View More Specs

Products 9
Product Code Brand Quantity Conjugate Form Concentration Price Quantity & Availability  
Product Code Brand Quantity Conjugate Form Concentration Price Quantity & Availability  
12652077
View Documents
Epredia
MS-704-B0
100 μL Biotin Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12672077
View Documents
Epredia
MS-704-B1
500 μL Biotin Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12662077
View Documents
Epredia
MS-704-B
1 mL Biotin Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12692077
View Documents
Epredia
MS-704-P0
100 μL Unconjugated Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12602087
View Documents
Epredia
MS-704-P1ABX
100 μL Unconjugated Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12622087
View Documents
Epredia
MS-704-PABX
200 μL Unconjugated Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12612087
View Documents
Epredia
MS-704-P1
500 μL Unconjugated Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12682077
View Documents
Epredia
MS-704-P
1 mL Unconjugated Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12662087
View Documents
Epredia
MS-704-R7
7 mL Unconjugated Prediluted Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Pulmonary surfactant is a complex mixture of phospholipids and proteins that is secreted from type II cells in alveoli and reduces the surface tension at the alveolar air-liquid interface, providing alveolar stability necessary for normal ventillation. Four distinct proteins isolated from pulmonary surfactant are termed surfactant proteins A, B, C, and D. SP-A (28-36kDa) and SP-D (43kDa) are collagenous carbohydrate-binding proteins, whereas SP-B (8-9kDa) and SP-C (4kDa) are non-collagenous hydrophobic proteins. SP-B is expressed in pulmonary adenocarcinomas with acinar, papillary, bronchioloalveolar, and solid growth patterns. Squamous cell and large cell carcinomas of the lung and nonpulmonary adenocarcinomas do not express SP-B. ProSP-B is glycosylated in the Golgi apparatus and undergoes carboxy- and amino-terminal proteolysis by a cathepsin D-like protease.

Host Species: Mouse

Clone: SPB01; same as 1-2-A5-9

Isotype: IgG2a/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant prosurfactant protein B

Molecular Weight: 42-46kDa

Positive Control: Lung adenocarcinoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Surfactant Protein B (Precursor) Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin)
Mouse
Cancer and Tumor Biology
Recombinant pro-surfactant protein B
Primary
SPB01; same as 1-2-A5-9
Monoclonal
RUO
Human, Rat
IgG2a κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.