Learn More
Epredia™ Lab Vision™ Oct-2 Rabbit Polyclonal Antibody with BSA and azide
Take advantage of a quality product that delivers accurate results with the Epredia™ Oct-2, Rabbit Polyclonal Antibody.
£146.00 - £550.80
Specifications
| Antigen | OCT2 |
|---|---|
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12633567
|
Epredia™
RB-9297-P0 |
100 μL | Concentrated | ||||||
|
12643567
|
Epredia™
RB-9297-P1 |
500 μL | Concentrated | ||||||
|
12623567
|
Epredia™
RB-9297-P |
1 mL | Concentrated | ||||||
|
12663567
|
Epredia™
RB-9297-R7 |
7 mL | Prediluted | ||||||
Description
Oct-2 is a transcription factor belonging to the POU homeodomain family that binds to the Ig gene octomer sites regulating B cell specific genes. Oct-2 protein expression is not restricted to B cells, although expression levels are much higher in these cells. Oct-2 expression is reported to be significantly greater in germinal center derived lymphomas, although other B cell lymphomas also display high levels of expression.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not tested.
Epitope: C-terminal
Immunogen: Synthetic peptide derived from C-terminus of human Oct-2
Molecular Weight: ∽80kDa
Positive Control: Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
| OCT2 | |
| Polyclonal | |
| Rabbit | |
| Transcription Regulation | |
| Synthetic peptide derived from C-terminus of human Oct-2 | |
| Primary |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.