missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Oct-2 Rabbit Polyclonal Antibody with BSA and azide

Take advantage of a quality product that delivers accurate results with the Epredia™ Oct-2, Rabbit Polyclonal Antibody.

Brand:  Epredia™ RB-9297-P1

 View more versions of this product

Product Code. 12643567

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Oct-2 is a transcription factor belonging to the POU homeodomain family that binds to the Ig gene octomer sites regulating B cell specific genes. Oct-2 protein expression is not restricted to B cells, although expression levels are much higher in these cells. Oct-2 expression is reported to be significantly greater in germinal center derived lymphomas, although other B cell lymphomas also display high levels of expression.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide derived from C-terminus of human Oct-2

Molecular Weight: ∽80kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

OCT2
Polyclonal
Unconjugated
Synthetic peptide derived from C-terminus of human Oct-2
RUO
Primary
Concentrated
Immunohistochemistry (Paraffin)
200µg/mL
Rabbit
500 μL
Transcription Regulation
Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.