missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Neurofilament (200kDa & 68kDa) Ab-1, Mouse Monoclonal Antibody

Take advantage of a quality product that delivers accurate results with the Epredia™ Neurofilament (200kDa & 68kDa) Ab-1, Mouse Monoclonal Antibody.

£112.00 - £349.86

Specifications

Antigen Neurofilament (200kDa and 68kDa) Ab-1
Clone 2F11
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Mehr Spezifikationen ansehen

Products 4
Produktcode Marke Quantity Form Preis Menge & Verfügbarkeit  
Produktcode Marke Quantity Form Preis Menge & Verfügbarkeit  
12651177
missing translation for 'viewDocuments'
Epredia™
MS-359-S0
100 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
12661177
missing translation for 'viewDocuments'
Epredia™
MS-359-S1
500 μL Concentrated
ONLINE EXCLUSIVE PRICE
£345.00
£316.63 / 500µL
Save £28.37
8% Off
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
12641177
missing translation for 'viewDocuments'
Epredia™
MS-359-S
1 mL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
12631177
missing translation for 'viewDocuments'
Epredia™
MS-359-R7
7 mL Prediluted
ONLINE EXCLUSIVE PRICE
£197.00
£185.05 / 7mL
Save £11.95
6% Off
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Beschreibung

Beschreibung

Neurofilaments are the intermediate filaments of neurons and their processes. They are protein triplets composed of three major subunits of MW 68kDa, 155kDa, and 200kDa. Neurofilaments are expressed in tumors of neural origin, or displaying neuronal differentiation, such as neuroblastoma, medulloblastoma, and retinoblastoma.

Host Species: Mouse

Clone: 2F11

Isotype: IgG1/κ

Species Reactivity: Human, Rabbit, Cat, Mouse and Rat. Does not react with dog. Others not known.

Epitope: Not determined

Immunogen: Neurofilament purified from human brain

Molecular Weight: 68 and 200kDa (phosphorylated forms of thsese subunits)

Positive Control: IMR-5 cells, Brain

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Spezifikation

Spezifikation

Neurofilament (200kDa and 68kDa) Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
IVD
Feline, Human, Mouse, Rabbit, Rat
IgG2a κ
2F11
Monoclonal
Mouse
Cytoskeletal Protein
Neurofilament purified from human brain
Primary
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt