missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Neurofilament (200kDa & 68kDa) Ab-1, Mouse Monoclonal Antibody

Take advantage of a quality product that delivers accurate results with the Epredia™ Neurofilament (200kDa & 68kDa) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-359-S0

 View more versions of this product

Product Code. 12651177

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Neurofilaments are the intermediate filaments of neurons and their processes. They are protein triplets composed of three major subunits of MW 68kDa, 155kDa, and 200kDa. Neurofilaments are expressed in tumors of neural origin, or displaying neuronal differentiation, such as neuroblastoma, medulloblastoma, and retinoblastoma.

Host Species: Mouse

Clone: 2F11

Isotype: IgG1/κ

Species Reactivity: Human, Rabbit, Cat, Mouse and Rat. Does not react with dog. Others not known.

Epitope: Not determined

Immunogen: Neurofilament purified from human brain

Molecular Weight: 68 and 200kDa (phosphorylated forms of thsese subunits)

Positive Control: IMR-5 cells, Brain

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Neurofilament (200kDa and 68kDa) Ab-1
Monoclonal
Unconjugated
Neurofilament purified from human brain
IVD
Primary
Concentrated
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
2F11
Mouse
100 μL
Cytoskeletal Protein
Feline, Human, Mouse, Rabbit, Rat
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.