Learn More
Insulin Receptor (α-Subunit) Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ Insulin Receptor (α-Subunit) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
£105.98 - £686.00
Specifications
| Antigen | Insulin Receptor (alpha-Subunit) Ab-1 |
|---|---|
| Clone | 83-14 |
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Form | Concentrated |
| Product Code | Brand | Quantity | Conjugate | Concentration | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Conjugate | Concentration | Price | Quantity & Availability | |||
|
12661827
|
Epredia
MS-631-B0 |
100 μL | Biotin | 200μg/mL | |||||
|
12671827
|
Epredia
MS-631-B1 |
500 μL | Biotin | 200μg/mL | |||||
|
12691827
|
Epredia
MS-631-P0 |
100 μL | Unconjugated | 200μg/mL | |||||
|
12601837
|
Epredia
MS-631-P1 |
500 μL | Unconjugated | 200μg/mL | |||||
|
12681827
|
Epredia
MS-631-P |
1 mL | Unconjugated | 200μg/mL | |||||
|
12611837
|
Epredia
MS-631-P1ABX |
100 μL | Unconjugated | 1mg/mL | |||||
|
12621837
|
Epredia
MS-631-PABX |
200 μL | Unconjugated | 1mg/mL | |||||
Description
The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain, whereas the α-subunit (135kDa) is completely extracellular.Host Species: Mouse
Clone: 83-14
Isotype: IgG2a/κ
Species Reactivity: Human. Weakly with Cow, Pig and Sheep. Does not react with rabbit, mouse and rat. Others not known.
Epitope: aa 469-592 (exon 7/8)
Immunogen: IM-9 lymphocytes followed by purified insulin receptor
Molecular Weight: 135kDa
Positive Control: IM-9 lymphocyte cells, Placenta
Cellular Localization: Membrane Ab-1 inhibits insulin binding and mimics insulin action. It is excellent for immunoprecipitation of alpha-subunit of IR.
Recommended for:
- Immunoprecipitation
- ELISA
- Insulin-like Agonist
- Inhibits (∽80%) Insulin Binding (10nM)
- Tyrosine Kinase Assay (Ab-mediated capture on microplates)
Note:
USA: RUO; Int'l: RUO
Specifications
| Insulin Receptor (alpha-Subunit) Ab-1 | |
| Immunoprecipitation | |
| Concentrated | |
| RUO | |
| Human | |
| IgG2a κ |
| 83-14 | |
| Monoclonal | |
| Mouse | |
| Steroids Hormones and Growth Factors | |
| IM-9 lymphocytes followed by purified insulin receptor | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.