missing translation for 'onlineSavingsMsg'
Learn More

Insulin Receptor (α-Subunit) Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ Insulin Receptor (α-Subunit) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-631-PABX

 View more versions of this product

Product Code. 12621837

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain, whereas the α-subunit (135kDa) is completely extracellular.

Host Species: Mouse

Clone: 83-14

Isotype: IgG2a/κ

Species Reactivity: Human. Weakly with Cow, Pig and Sheep. Does not react with rabbit, mouse and rat. Others not known.

Epitope: aa 469-592 (exon 7/8)

Immunogen: IM-9 lymphocytes followed by purified insulin receptor

Molecular Weight: 135kDa

Positive Control: IM-9 lymphocyte cells, Placenta

Cellular Localization: Membrane Ab-1 inhibits insulin binding and mimics insulin action. It is excellent for immunoprecipitation of alpha-subunit of IR.

Recommended for:

  • Immunoprecipitation
  • ELISA
  • Insulin-like Agonist
  • Inhibits (∽80%) Insulin Binding (10nM)
  • Tyrosine Kinase Assay (Ab-mediated capture on microplates)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Insulin Receptor (alpha-Subunit) Ab-1
Monoclonal
1mg/mL
Mouse
200 μL
Steroids Hormones and Growth Factors
Human
IgG2a κ
Immunoprecipitation
83-14
Unconjugated
IM-9 lymphocytes followed by purified insulin receptor
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.