Learn More
Epredia™ Lab Vision™ HER-4/c-erbB-4 Rabbit Polyclonal Antibody with BSA and azide
Recommended for Immunohistochemistry (Formalin/paraffin) and Western Blotting, Epredia™ HER-4/c-erbB-4, Rabbit Polyclonal Antibody provides accurate, reproducible results.
£364.14
Specifications
| Antigen | HER-4/c-erbB-4 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12603417
|
Epredia™
RB-9045-P1 |
500 μL | Concentrated | ||||||
Description
c-erbB-4 is the fourth member of class I receptor kinase family. HER-4 is most predominantly expressed in several breast carcinoma cell lines, and in normal skeletal muscle, heart, pituitary, brain, and cerebellum. Breast tumor cell lines T47-D, MDA-MB-453, BT-474 and H3396 are found to have the highest levels of mRNA, and intermediate levels are seen in MCF-7, MDA-MB-330 and MDA-MB-361. Expression of erbB-4 is low or absent in some breast tumor cell lines such as MDA-MB-231, MDA-MB-157, MDA-MB-468, and SKBR-3.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Mouse. Others not tested.
Epitope: C-terminal
Immunogen: A synthetic peptide from C-terminus of human HER-4 protein
Molecular Weight: 185kDa
Positive Control: Breast CA T47D cells
Cellular Localization: Cytoplasmic and membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
| HER-4/c-erbB-4 | |
| Polyclonal | |
| Rabbit | |
| EGFR Family | |
| A synthetic peptide from C-terminus of human c-erbB-4 protein | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.