missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ HER-4/c-erbB-4 Rabbit Polyclonal Antibody with BSA and azide

Recommended for Immunohistochemistry (Formalin/paraffin) and Western Blotting, Epredia™ HER-4/c-erbB-4, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Brand:  Epredia™ RB-9045-P1

 View more versions of this product

Product Code. 12603417

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

c-erbB-4 is the fourth member of class I receptor kinase family. HER-4 is most predominantly expressed in several breast carcinoma cell lines, and in normal skeletal muscle, heart, pituitary, brain, and cerebellum. Breast tumor cell lines T47-D, MDA-MB-453, BT-474 and H3396 are found to have the highest levels of mRNA, and intermediate levels are seen in MCF-7, MDA-MB-330 and MDA-MB-361. Expression of erbB-4 is low or absent in some breast tumor cell lines such as MDA-MB-231, MDA-MB-157, MDA-MB-468, and SKBR-3.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of human HER-4 protein

Molecular Weight: 185kDa

Positive Control: Breast CA T47D cells

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HER-4/c-erbB-4
Polyclonal
Unconjugated
A synthetic peptide from C-terminus of human c-erbB-4 protein
RUO
Primary
Concentrated
Western Blot
200µg/mL
Rabbit
500 μL
EGFR Family
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.