missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ HER-2/c-erbB-2/neu Rabbit Monoclonal Antibody supernatant

Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ HER-2/c-erbB-2/neu, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Brand:  Epredia™ RM-2112-S0

 View more versions of this product

Product Code. 12653647

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

HER2 gene encodes a transmembrane glycoprotein with tyrosine kinase activity. It is closely related in structure to the epidermal growth factor receptor c-erbB-3/HER3/neu oncoprotein. The protein is detectable in a subset of breast and other carcinomas (urinary bladder, stomach, lung, endometrium and ovary). In the case of breast cancer, over expression determined by immunohistochemistry has been shown to be associated with poor prognosis and an indication to initiate trastuzumab/herceptin therapy.

Host Species: Rabbit

Clone: EP1045Y

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: A synthetic peptide corresponding to the c-terminus of human Her2

Molecular Weight: 185 kDa

Positive Control: Breast carcinomas

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HER-2/c-erbB-2/neu
EP1045Y
Rabbit
100 μL
Human
Immunohistochemistry
Unconjugated
A synthetic peptide corresponding to the c-terminus of human Her2
Primary
Supernatant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.