missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ HER-2/c-erbB-2/neu (Phospho-specific) Ab-18 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Provide accurate, reproducible results with the Epredia™ HER-2/c-erbB-2/neu (Phospho-specific) Ab-18, Mouse Monoclonal Antibody.

£183.90 - £904.00

Specifications

Antigen HER-2/c-erbB-2/neu (Phospho-specific) Ab-18
Clone PN2A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Form Concentrated
View More Specs

Products 8
Product Code Brand Quantity Conjugate Concentration Price Quantity & Availability  
Product Code Brand Quantity Conjugate Concentration Price Quantity & Availability  
12658846
View Documents
Epredia™
MS-1072-B0
100 μL Biotin 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12668846
View Documents
Epredia™
MS-1072-B1
500 μL Biotin 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12648846
View Documents
Epredia™
MS-1072-B
1 mL Biotin 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12688846
missing translation for 'viewDocuments'
Epredia™
MS-1072-P0
100 μL Unconjugated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
12698846
missing translation for 'viewDocuments'
Epredia™
MS-1072-P1
500 μL Unconjugated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
12678846
missing translation for 'viewDocuments'
Epredia™
MS-1072-P
1 mL Unconjugated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
12608856
missing translation for 'viewDocuments'
Epredia™
MS-1072-P1ABX
100 μL Unconjugated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
12618856
missing translation for 'viewDocuments'
Epredia™
MS-1072-PABX
200 μL Unconjugated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Beschreibung

Beschreibung

c-erbB-2 (HER-2/neu) is a 185 kDa receptor tyrosine kinase belonging to the epidermal growth factor receptor family, which also includes EGFR (HER-1), c-erbB-3 (HER-3), and c-erbB-4 (HER-4). Amplification of the HER-2 gene with overexpression of the protein occurs in 20-30% of breast cancers, and tumor progression in mammary cancer is associated with elevated levels of tyrosine-phosphorylated neu and erbB-3. Tyr1248 is the phosphorylation site that is most tightly linked to oncogenic transformation and coupling to the ras/MAP kinase signaling pathway. Cases of invasive breast carcinoma with the phosphorylated receptor have displayed aggressive clinicopathological features and adverse prognoses. Activation of HER-2 is frequent in ductal carcinoma in situ suggesting that HER-2 signaling plays a critical role in the early stages of breast tumorigenesis.

Host Species: Mouse

Clone: PN2A

Isotype: IgG1

Species Reactivity: Human and Rat. Others not tested.

Epitope: aa 1242-1255 with pTyr

Immunogen: Tyrosine-phosphorylated synthetic peptide corresponding to aa 1242-1255 [Lys-TAENPE-pY-LGLDVPV] from the C-terminus of human HER-2 protein (P-Tyr1248)

Molecular Weight: 185 kDa

Positive Control: EGF-treated SKBR-3 cells. About 5% of breast CA are positive for phospho-HER-2/c-erbB-2/neu oncoprotein.

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Not verified)
  • Flow Cytometry (Not verified)

Note:

USA: RUO; Int'l: RUO

Spezifikation

Spezifikation

HER-2/c-erbB-2/neu (Phospho-specific) Ab-18
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Concentrated
RUO
Human, Rat
IgG1
PN2A
Monoclonal
Mouse
EGFR Family
Tyrosine-phosphorylated synthetic peptide corresponding to aa1242-1255 [Lys-TAENPE-pY-LGLDVPV] from the C-terminus of human c-erbB-2 protein (P-Tyr1248)
Primary
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt