missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ HER-2/c-erbB-2/neu (Phospho-specific) Ab-18 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Provide accurate, reproducible results with the Epredia™ HER-2/c-erbB-2/neu (Phospho-specific) Ab-18, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1072-B

 View more versions of this product

Product Code. 12648846

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

c-erbB-2 (HER-2/neu) is a 185 kDa receptor tyrosine kinase belonging to the epidermal growth factor receptor family, which also includes EGFR (HER-1), c-erbB-3 (HER-3), and c-erbB-4 (HER-4). Amplification of the HER-2 gene with overexpression of the protein occurs in 20-30% of breast cancers, and tumor progression in mammary cancer is associated with elevated levels of tyrosine-phosphorylated neu and erbB-3. Tyr1248 is the phosphorylation site that is most tightly linked to oncogenic transformation and coupling to the ras/MAP kinase signaling pathway. Cases of invasive breast carcinoma with the phosphorylated receptor have displayed aggressive clinicopathological features and adverse prognoses. Activation of HER-2 is frequent in ductal carcinoma in situ suggesting that HER-2 signaling plays a critical role in the early stages of breast tumorigenesis.

Host Species: Mouse

Clone: PN2A

Isotype: IgG1

Species Reactivity: Human and Rat. Others not tested.

Epitope: aa 1242-1255 with pTyr

Immunogen: Tyrosine-phosphorylated synthetic peptide corresponding to aa 1242-1255 [Lys-TAENPE-pY-LGLDVPV] from the C-terminus of human HER-2 protein (P-Tyr1248)

Molecular Weight: 185 kDa

Positive Control: EGF-treated SKBR-3 cells. About 5% of breast CA are positive for phospho-HER-2/c-erbB-2/neu oncoprotein.

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Not verified)
  • Flow Cytometry (Not verified)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HER-2/c-erbB-2/neu (Phospho-specific) Ab-18
Monoclonal
200μg/mL
Mouse
1 mL
EGFR Family
Human, Rat
IgG1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
PN2A
Biotin
Tyrosine-phosphorylated synthetic peptide corresponding to aa1242-1255 [Lys-TAENPE-pY-LGLDVPV] from the C-terminus of human c-erbB-2 protein (P-Tyr1248)
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.