missing translation for 'onlineSavingsMsg'
Learn More

HER-2/c-erbB-2/neu Ab-16 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ HER-2/c-erbB-2/neu Ab-16, Mouse Monoclonal Antibody.

£105.98 - £615.60

Specifications

Antigen HER-2/c-erbB-2/neu Ab-16
Clone L26
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Product Code Brand Quantity Concentration Price Quantity & Availability  
Product Code Brand Quantity Concentration Price Quantity & Availability  
12611737
View Documents
Epredia
MS-600-P0A
100 μL 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12601737
View Documents
Epredia
MS-600-P1A
500 μL 200μg/mL
ONLINE EXCLUSIVE PRICE
£390.00
£366.79 / 500µL
Save £23.21
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12621737
View Documents
Epredia
MS-600-P1ABX
100 μL 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12641737
View Documents
Epredia
MS-600-PABX
200 μL 1mg/mL
ONLINE EXCLUSIVE PRICE
£658.00
£615.60 / 200µL
Save £42.40
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Mouse

Clone: L26

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Involved in the formation of heterodimers

Immunogen: Extracellular domain of recombinant human HER-2/c-erbB-2 oncoprotein

Molecular Weight: 185kDa

Positive Control: SKBR-3 cells

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Not suitable)
  • Immunoprecipitation (Native only)
  • Inhibits Binding of EGF (∽70%), NDF (∽40%)
  • Reduces Affinity of EGF (3-fold) and NDF (2-fold)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

HER-2/c-erbB-2/neu Ab-16
Immunoprecipitation
Unconjugated
Mouse
EGFR Family
Extracellular domain of recombinant human c-erbB-2/HER-2 oncoprotein
Primary
L26
Monoclonal
Concentrated
RUO
Human
IgG2a κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.