missing translation for 'onlineSavingsMsg'
Learn More

HER-2/c-erbB-2/neu Ab-16 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ HER-2/c-erbB-2/neu Ab-16, Mouse Monoclonal Antibody.

Brand:  Epredia MS-600-P0A

 View more versions of this product

Product Code. 12611737

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Mouse

Clone: L26

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Involved in the formation of heterodimers

Immunogen: Extracellular domain of recombinant human HER-2/c-erbB-2 oncoprotein

Molecular Weight: 185kDa

Positive Control: SKBR-3 cells

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Not suitable)
  • Immunoprecipitation (Native only)
  • Inhibits Binding of EGF (∽70%), NDF (∽40%)
  • Reduces Affinity of EGF (3-fold) and NDF (2-fold)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HER-2/c-erbB-2/neu Ab-16
Monoclonal
200μg/mL
Mouse
100 μL
EGFR Family
Human
IgG2a κ
Immunoprecipitation
L26
Unconjugated
Extracellular domain of recombinant human c-erbB-2/HER-2 oncoprotein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Dokument

Dokument

Produktspecifikationer
Kampanjer

Kampanjer

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.