Learn More
HER-2/c-erbB-2/neu Ab-1 Rabbit Polyclonal Antibody, Epredia™
Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation, Epredia™ HER-2/c-erbB-2/neu Ab-1, Rabbit Polyclonal Antibody provides accurate, reproducible results.
£129.54 - £639.00
Specifications
| Antigen | HER-2/c-erbB-2/neu Ab-1 |
|---|---|
| Concentration | 1mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Purification Method | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Purification Method | Price | Quantity & Availability | ||||
|
12602797
|
Epredia
RB-103-P0 |
100 μL | Purified with BSA and Azide | ||||||
|
12612797
|
Epredia
RB-103-P1 |
500 μL | Purified with BSA and Azide | ||||||
|
12622797
|
Epredia
RB-103-P1ABX |
500 μL | Purified w/o BSA and Azide | ||||||
|
12692787
|
Epredia
RB-103-P |
1 mL | Purified with BSA and Azide | ||||||
|
12632797
|
Epredia
RB-103-PABX |
1 mL | Purified w/o BSA and Azide | ||||||
Description
c-erbB-2, second member (c-erbB-2/HER-2/neu) of the c-erbB family is a receptor tyrosine kinase. It exhibits extracellular domains with two cysteine-rich sequences, and a cytoplasmic tyrosine kinase domain flanked by large hydrophilic tails that carry several tyrosine autophosphorylation sites. Approximately 25% of primary breast and ovarian tumors were found to overexpress the proteinHost Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Monkey, Rat and Mouse. Others not tested.
Epitope: C-terminal
Immunogen: Synthetic peptide derived from C-terminus of human HER-2/c-erbB-2 protein. This sequence is identical in rat neu protein.
Molecular Weight: 185 kDa
Positive Control: SKBR-3 cells, or T47D cells, or Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
| HER-2/c-erbB-2/neu Ab-1 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Oncogenesis | |
| Synthetic peptide derived from C-terminus of human c-erbB-2/HER-2 protein. This Sequence is identical in rat neu protein. | |
| Primary |
| 1mg/mL | |
| Polyclonal | |
| Concentrated | |
| RUO | |
| Human, Mouse, Non-human Primate | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.