missing translation for 'onlineSavingsMsg'
Learn More

HER-2/c-erbB-2/neu Ab-1 Rabbit Polyclonal Antibody, Epredia™

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation, Epredia™ HER-2/c-erbB-2/neu Ab-1, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Brand:  Epredia RB-103-P1ABX

 View more versions of this product

Product Code. 12622797

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

c-erbB-2, second member (c-erbB-2/HER-2/neu) of the c-erbB family is a receptor tyrosine kinase. It exhibits extracellular domains with two cysteine-rich sequences, and a cytoplasmic tyrosine kinase domain flanked by large hydrophilic tails that carry several tyrosine autophosphorylation sites. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Monkey, Rat and Mouse. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide derived from C-terminus of human HER-2/c-erbB-2 protein. This sequence is identical in rat neu protein.

Molecular Weight: 185 kDa

Positive Control: SKBR-3 cells, or T47D cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HER-2/c-erbB-2/neu Ab-1
Polyclonal
Unconjugated
Synthetic peptide derived from C-terminus of human c-erbB-2/HER-2 protein. This Sequence is identical in rat neu protein.
500 μL
Oncogenesis
Human, Mouse, Non-human Primate
IgG
Immunoprecipitation, Western Blot
1mg/mL
Rabbit
Purified w/o BSA and Azide
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.