Learn More
Growth Hormone, Human/hGH, Ab-3 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments.
£104.04 - £444.00
Specifications
| Antigen | Growth Hormone, Human/hGH Ab-3 |
|---|---|
| Clone | GH01 / #2 + GH02 / #7 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Form | Concentration | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Concentration | Price | Quantity & Availability | |||
|
12669566
|
Epredia
MS-1328-P0 |
100 μL | Concentrated | 200μg/mL | |||||
|
12679566
|
Epredia
MS-1328-P1 |
500 μL | Concentrated | 200μg/mL |
£387.00 £320.16 / 500µL Save £66.84 17% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
|
12639566
|
Epredia
MS-1328-P |
1 mL | Concentrated | 200μg/mL | |||||
|
12629576
|
Epredia
MS-1328-R7 |
7 mL | Prediluted | Prediluted | |||||
Description
hGH is produced by the somatotroph cells in the pituitary. hGH acts through binding to membrane receptors that belong to the cytokine receptor superfamily. Ligand binding induces receptor dimerization and activation of the receptor-associated kinase: JAK2; this results in phosphorylation of the kinase itself, of the receptor, and of many cellular proteins including the Stat proteins as well as adaptors leading to the activation of the Ras/MAP kinase pathway and of the PI-3 kinase pathway.Host Species: Mouse
Clone: GH01/#2 + GH02/#7
Isotype: IgG2a/κ + IgG2a/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Purified recombinant human growth hormone
Molecular Weight: ∽22 kDa
Positive Control: Recombinant human growth hormone, Human anterior pituitary
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
| Growth Hormone, Human/hGH Ab-3 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| IgG2a κ |
| GH01 / #2 + GH02 / #7 | |
| Monoclonal | |
| Mouse | |
| Steroids Hormones and Growth Factors | |
| Purified recombinant human growth hormone | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.