missing translation for 'onlineSavingsMsg'
Learn More

Growth Hormone, Human/hGH, Ab-3 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments.

Brand:  Epredia MS-1328-P

 View more versions of this product

Product Code. 12639566

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

hGH is produced by the somatotroph cells in the pituitary. hGH acts through binding to membrane receptors that belong to the cytokine receptor superfamily. Ligand binding induces receptor dimerization and activation of the receptor-associated kinase: JAK2; this results in phosphorylation of the kinase itself, of the receptor, and of many cellular proteins including the Stat proteins as well as adaptors leading to the activation of the Ras/MAP kinase pathway and of the PI-3 kinase pathway.

Host Species: Mouse

Clone: GH01/#2 + GH02/#7

Isotype: IgG2a/κ + IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant human growth hormone

Molecular Weight: ∽22 kDa

Positive Control: Recombinant human growth hormone, Human anterior pituitary

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Growth Hormone, Human/hGH Ab-3
Monoclonal
200μg/mL
Mouse
1 mL
Steroids Hormones and Growth Factors
Human
IgG2a κ
Immunohistochemistry (Paraffin), Western Blot
GH01 / #2 + GH02 / #7
Unconjugated
Purified recombinant human growth hormone
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.