missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Estrogen Receptor Ab-10 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Ensure accurate, reproducible results in western blotting, immunoprecipitation, gel supershift and chromatin precipitation experiments.

£116.28 - £525.30

Specifications

Antigen Estrogen Receptor Ab-10
Clone TE111.5D11
Applications Immunoprecipitation
Classification Monoclonal
Form Concentrated
View More Specs

Products 6
Product Code Brand Quantity Conjugate Concentration Price Quantity & Availability  
Product Code Brand Quantity Conjugate Concentration Price Quantity & Availability  
12650967
View Documents
Epredia™
MS-315-B0
100 μL Biotin 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12660967
View Documents
Epredia™
MS-315-B1
500 μL Biotin 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12680967
View Documents
Epredia™
MS-315-P0
100 μL Unconjugated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12690967
View Documents
Epredia™
MS-315-P1
500 μL Unconjugated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12670967
View Documents
Epredia™
MS-315-P
1 mL Unconjugated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12610977
View Documents
Epredia™
MS-315-PABX
200 μL Unconjugated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

The ER gene consists of more than 140kb of genomic DNA divided into 8 exons and is translated into a protein with six functionally discrete domains required for transcription activation function, binding to estrogen response element (ERE) constitutive dimerization, binding to heat shock proteins, and ligand recognition. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Host Species: Mouse

Clone: TE111.5D11

Isotype: IgG1

Species Reactivity: Human, Cow, Pig, Dog, Mouse, Rat, Sheep, Rabbit, Hamster and Chicken. Others not known.

Epitope: aa 302-553

Immunogen: 35 kDa, C-terminal fragment (aa 302-595) of human ER expressed in E. coli

Molecular Weight: 67 kDa

Positive Control: T47D cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native and denatured)
  • Gel Supershift
  • Blocks Binding of Estrogen to ER
  • Chromatin Precipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Estrogen Receptor Ab-10
Immunoprecipitation
Concentrated
RUO
Bovine, Canine, Chicken, Hamster, Human, Mouse, Pig, Rabbit, Rat, Sheep
IgG1
TE111.5D11
Monoclonal
Mouse
Steroids Hormones and Growth Factors
35kDa, C-terminal fragment (aa 302-595) of human ER expressed in E. coli
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.