missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Estrogen Receptor Ab-10 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Ensure accurate, reproducible results in western blotting, immunoprecipitation, gel supershift and chromatin precipitation experiments.

Brand:  Epredia™ MS-315-P

 View more versions of this product

Product Code. 12670967

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

The ER gene consists of more than 140kb of genomic DNA divided into 8 exons and is translated into a protein with six functionally discrete domains required for transcription activation function, binding to estrogen response element (ERE) constitutive dimerization, binding to heat shock proteins, and ligand recognition. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Host Species: Mouse

Clone: TE111.5D11

Isotype: IgG1

Species Reactivity: Human, Cow, Pig, Dog, Mouse, Rat, Sheep, Rabbit, Hamster and Chicken. Others not known.

Epitope: aa 302-553

Immunogen: 35 kDa, C-terminal fragment (aa 302-595) of human ER expressed in E. coli

Molecular Weight: 67 kDa

Positive Control: T47D cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native and denatured)
  • Gel Supershift
  • Blocks Binding of Estrogen to ER
  • Chromatin Precipitation

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Estrogen Receptor Ab-10
Monoclonal
200μg/mL
Mouse
1 mL
Steroids Hormones and Growth Factors
Bovine, Canine, Chicken, Hamster, Human, Mouse, Pig, Rabbit, Rat, Sheep
IgG1
Immunoprecipitation
TE111.5D11
Unconjugated
35kDa, C-terminal fragment (aa 302-595) of human ER expressed in E. coli
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.