Learn More
Epredia™ Lab Vision™ Epstein-Barr Virus/LMP Ab-1 Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epstein-Barr Virus/LMP Ab-1, Mouse Monoclonal Antibody.
£97.82 - £390.66
Specifications
| Antigen | Epstein-Barr Virus/LMP Ab-1 |
|---|---|
| Clone | CS1+CS2+CS3+CS4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12699856
|
Epredia™
MS-1458-S0 |
100 μL | Concentrated | ||||||
|
12609866
|
Epredia™
MS-1458-S1 |
500 μL | Concentrated | ||||||
|
12689856
|
Epredia™
MS-1458-S |
1 mL | Concentrated | ||||||
|
12679856
|
Epredia™
MS-1458-R7 |
7 mL | Prediluted | ||||||
Description
Host Species: Mouse
Clone: CS1 + CS2 + CS3 + CS4
Isotype: IgG1/κ
Species Reactivity: EBV-positive cells
Epitope: Located in the internal part of the membrane-associated viral LMP
Immunogen: EBV-encoded recombinant latent membrane protein
Molecular Weight: 60 kDa (LMP)
Positive Control: EBV-positive lymphoblastoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| Epstein-Barr Virus/LMP Ab-1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| ASR, IVD | |
| EBV-encoded recombinant latent membrane protein | |
| Primary |
| CS1+CS2+CS3+CS4 | |
| Monoclonal | |
| Mouse | |
| Infectious Disease | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.