Learn More
Epredia™ Lab Vision™ Epstein-Barr Virus/LMP Ab-1 Mouse Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: CS1 + CS2 + CS3 + CS4
Isotype: IgG1/κ
Species Reactivity: EBV-positive cells
Epitope: Located in the internal part of the membrane-associated viral LMP
Immunogen: EBV-encoded recombinant latent membrane protein
Molecular Weight: 60 kDa (LMP)
Positive Control: EBV-positive lymphoblastoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Epstein-Barr Virus/LMP Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CS1+CS2+CS3+CS4 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | EBV-encoded recombinant latent membrane protein |
| Quantity | 500 μL |
| Regulatory Status | ASR, IVD |
| Research Discipline | Infectious Disease |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.