missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Epithelial Specific Antigen/Ep-CAM Ab-9, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Specific Antigen/Ep-CAM Ab-9, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1898-R7

Code : LD

Additional Details : Weight : 0.01200kg

 View more versions of this product

Product Code. 12670417

  • £237.00 / 7mL
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Host Species: Mouse

Clone: Ber-EP4

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: MCF-7 human breast carcinoma cell line

Molecular Weight: ∽34 and 39 kDa (reducing); 39 and 41 kDa (nonreducing); 31 and 36 kDa (deglycosylated)

Positive Control: Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Epithelial Specific Antigen/Ep-CAM Ab-9
Monoclonal
Unconjugated
MCF-7 human breast carcinoma cell line
IVD
Primary
Prediluted
Immunohistochemistry (Paraffin)
Ber-EP4
Mouse
7 mL
Cancer and Tumor Biology
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers