Learn More
Epredia™ Lab Vision™ Epithelial Specific Antigen/Ep-CAM Ab-9, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Specific Antigen/Ep-CAM Ab-9, Mouse Monoclonal Antibody.
£115.00 - £474.32
Tekniske data
| Antigen | Epithelial Specific Antigen/Ep-CAM Ab-9 |
|---|---|
| Clone | Ber-EP4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Produktkode | Brand | Quantity | Form | Pris | Antal & tilgængelighed | ||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Form | Pris | Antal & tilgængelighed | ||||
|
12600427
|
Epredia™
MS-1898-S0 |
100 μL | Concentrated | ||||||
|
12610427
|
Epredia™
MS-1898-S1 |
500 μL | Concentrated |
£355.00 £334.03 / 500µL Save £20.97 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12690417
|
Epredia™
MS-1898-S |
1 mL | Concentrated |
£503.00 £474.32 / 1mL Save £28.68 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12670417
|
Epredia™
MS-1898-R7 |
7 mL | Prediluted |
£246.00 £219.29 / 7mL Save £26.71 10% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beskrivelse
Host Species: Mouse
Clone: Ber-EP4
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: MCF-7 human breast carcinoma cell line
Molecular Weight: ∽34 and 39 kDa (reducing); 39 and 41 kDa (nonreducing); 31 and 36 kDa (deglycosylated)
Positive Control: Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Tekniske data
| Epithelial Specific Antigen/Ep-CAM Ab-9 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Human | |
| IgG2a κ |
| Ber-EP4 | |
| Monoclonal | |
| Mouse | |
| Cancer and Tumor Biology | |
| MCF-7 human breast carcinoma cell line | |
| Primary |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.