Learn More
Epredia™ Lab Vision™ Epithelial Specific Antigen/Ep-CAM Ab-9, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Specific Antigen/Ep-CAM Ab-9, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1898-S1
Code : LD
Additional Details : Weight : 0.00190kg
Description
Host Species: Mouse
Clone: Ber-EP4
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: MCF-7 human breast carcinoma cell line
Molecular Weight: ∽34 and 39 kDa (reducing); 39 and 41 kDa (nonreducing); 31 and 36 kDa (deglycosylated)
Positive Control: Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Epithelial Specific Antigen/Ep-CAM Ab-9 | |
Monoclonal | |
Unconjugated | |
MCF-7 human breast carcinoma cell line | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
Ber-EP4 | |
Mouse | |
500 μL | |
Cancer and Tumor Biology | |
Human | |
IgG2a κ |