missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ DOG1 Rabbit Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Dog1, Rabbit Monoclonal Antibody, part of a comprehensive antibody portfolio.

£222.00 - £1240.00

Specifications

Antigen DOG1
Clone SP31
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12673797
View Documents
Epredia™
RM-9132-S0
100 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12663797
View Documents
Epredia™
RM-9132-S
1 mL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12643797
View Documents
Epredia™
RM-9132-R7
7 mL Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12653797
View Documents
Epredia™
RM-9132-RQ
12 mL Ready-to-Use This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

DOG1 is a cell surface protein of unknown function selectively expressed in GIST. Among GIST cases with KIT mutations the DOG1 antibody identified 11% more cases than c-Kit antibody. DOG1 identifies vast majority of both c-Kit negative and PDGFRA mutated GIST cases that may still benefit from imatinib mesylate (Gleevac), an inhibitor of the kit tyrosin kinase. In addition, DOG1 immunoreactivity is seen in fewer cases of mesanchymal, epithelial tumors and melanomas when compared with c-Kit. The use of this highly sensitive and specific novel marker will increase the accuracy of GIST diagnosis.

Host Species: Rabbit

Clone: SP31

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: Synthetic peptide of human DOG1

Molecular Weight: 78 kDa

Positive Control: Gastrointestinal Stromal Tumors

Cellular Localization: Cytoplasmic and Membranous

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

DOG1
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Synthetic peptide of human Dog1
SP31
Monoclonal
Rabbit
Cancer and Tumor Biology
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.