missing translation for 'onlineSavingsMsg'
Learn More

CD99/MIC2 (Ewing’s Sarcoma Marker) Ab-2 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry applications with Epredia™ CD99/MIC2 (Ewing's Sarcoma Marker) Ab-2, Mouse Monoclonal Antibody.

£104.04 - £449.89

Specifica

Antigen CD99/MIC2 (Ewings Sarcoma Marker) Ab-2
Clone O13
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
Visualizza altre specifiche

Prodotti 6
Codice del prodotto Marca Quantity Form Concentration Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantity Form Concentration Prezzo Quantità e disponibilità  
12630017
missing translation for 'viewDocuments'
Epredia
MS-1633-P0
100 μL Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
12640017
missing translation for 'viewDocuments'
Epredia
MS-1633-P1ABX
100 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
12650017
missing translation for 'viewDocuments'
Epredia
MS-1633-PABX
200 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
12620017
missing translation for 'viewDocuments'
Epredia
MS-1633-P1
500 μL Concentrated 200μg/mL
ONLINE EXCLUSIVE PRICE
£355.00
£334.47 / 500µL
Save £20.53
5% Off
Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
12610017
missing translation for 'viewDocuments'
Epredia
MS-1633-P
1 mL Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
12690017
missing translation for 'viewDocuments'
Epredia
MS-1633-R7
7 mL Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
Descrizione

Descrizione

CD99, or MIC2 gene product, or E2 antigen is expressed on the cell membrane of some lymphocytes, cortical thymocytes, and granulosa cells of the ovary. The antigen is also expressed by most pancreatic islet cells, Sertoli cells of the testis, and some endothelial cells. Mature granulocytes express very little or no CD99. MIC2 is strongly expressed on Ewing's sarcoma cells and primitive peripheral neuroectodermal tumors.

Host Species: Mouse

Clone: O13

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Not determined

Immunogen: MeWo human melanoma cells

Molecular Weight: 27-32 kDa

Positive Control: Pancreas or Ewing's sarcoma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifica

Specifica

CD99/MIC2 (Ewings Sarcoma Marker) Ab-2
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
IgG1
O13
Monoclonal
Mouse
Oncogenesis
MeWo human melanoma cells
Primary
Videos
SDS
Documenti

Documenti

Certificati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato