missing translation for 'onlineSavingsMsg'
Learn More

CD99/MIC2 (Ewing’s Sarcoma Marker) Ab-2 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry applications with Epredia™ CD99/MIC2 (Ewing's Sarcoma Marker) Ab-2, Mouse Monoclonal Antibody.

Brand:  Epredia MS-1633-R7

 View more versions of this product

Product Code. 12690017

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

CD99, or MIC2 gene product, or E2 antigen is expressed on the cell membrane of some lymphocytes, cortical thymocytes, and granulosa cells of the ovary. The antigen is also expressed by most pancreatic islet cells, Sertoli cells of the testis, and some endothelial cells. Mature granulocytes express very little or no CD99. MIC2 is strongly expressed on Ewing's sarcoma cells and primitive peripheral neuroectodermal tumors.

Host Species: Mouse

Clone: O13

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Not determined

Immunogen: MeWo human melanoma cells

Molecular Weight: 27-32 kDa

Positive Control: Pancreas or Ewing's sarcoma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD99/MIC2 (Ewings Sarcoma Marker) Ab-2
Monoclonal
Unconjugated
MeWo human melanoma cells
RUO
Primary
Prediluted
Immunohistochemistry (Paraffin)
O13
Mouse
7 mL
Oncogenesis
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.