Learn More
Epredia™ Lab Vision™ CD68/Macrophage Marker Ab-4, Mouse Monoclonal Antibody
Test for macrophages and monocytes with Epredia™ CD68/Macrophage Marker Ab-4, Mouse Monoclonal Antibody.
£90.90 - £407.01
Specifications
| Antigen | CD68/Macrophage Marker Ab-4 |
|---|---|
| Clone | PG-M1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12630167
|
Epredia™
MS-1808-S0 |
100 μL | Concentrated | ||||||
|
12640167
|
Epredia™
MS-1808-S1 |
500 μL | Concentrated |
£315.00 £296.15 / 500µL Save £18.85 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12610167
|
Epredia™
MS-1808-S |
1 mL | Concentrated |
£518.00 £407.01 / 1mL Save £110.99 21% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12600167
|
Epredia™
MS-1808-R7 |
7 mL | Prediluted |
£307.00
7mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CD68 glycoprotein is expressed on macrophages and monocytes.Host Species: Mouse
Clone: PG-M1
Isotype: IgG3/κ
Species Reactivity: Human. Others not tested
Epitope: Not determined
Immunogen: Gaucher cells
Molecular Weight: 110 kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| CD68/Macrophage Marker Ab-4 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Human | |
| IgG3 κ |
| PG-M1 | |
| Monoclonal | |
| Mouse | |
| Hematopoietic | |
| Gaucher cells | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.