Learn More
Epredia™ Lab Vision™ CD68/Macrophage Marker Ab-4, Mouse Monoclonal Antibody
Test for macrophages and monocytes with Epredia™ CD68/Macrophage Marker Ab-4, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1808-R7
Description
CD68 glycoprotein is expressed on macrophages and monocytes.Host Species: Mouse
Clone: PG-M1
Isotype: IgG3/κ
Species Reactivity: Human. Others not tested
Epitope: Not determined
Immunogen: Gaucher cells
Molecular Weight: 110 kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| CD68/Macrophage Marker Ab-4 | |
| Monoclonal | |
| Unconjugated | |
| Gaucher cells | |
| IVD | |
| Primary | |
| Prediluted |
| Immunohistochemistry (Paraffin) | |
| PG-M1 | |
| Mouse | |
| 7 mL | |
| Hematopoietic | |
| Human | |
| IgG3 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.