Learn More
Epredia™ Lab Vision™ CD2 (LFA-2) Ab-2, Mouse Monoclonal Antibody
Obtain accurate and reproducible results in immunohistochemistry experiments with Epredia™ CD2 (LFA-2) Ab-2, Mouse Monoclonal Antibody.
£156.06 - £552.84
Specifications
| Antigen | CD2 (LFA-2) Ab-2 |
|---|---|
| Clone | AB75 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12658856
|
Epredia™
MS-1074-S0 |
100 μL | Concentrated | ||||||
|
12678856
|
Epredia™
MS-1074-S1 |
500 μL | Concentrated |
£416.00 £391.19 / 500µL Save £24.81 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12668856
|
Epredia™
MS-1074-S |
1 mL | Concentrated |
£591.00 £552.84 / 1mL Save £38.16 6% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12648856
|
Epredia™
MS-1074-R7 |
7 mL | Prediluted |
£397.00 £373.97 / 7mL Save £23.03 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CD2, which is also known as E-rosette receptor, T11 and lymphocyte-function antigen-2 (LFA-2), is expressed on T cells, thymocytes, and subset of natural killer cells. Human CD2 functions as the receptor for sheep erythrocytes, human CD58 (LFA-3), and CD15s (Sialyl Lewis X). p56lck, p59fyn, CD3η and CD3ε are tyrosine-phosphorylated after CD2 stimulation. CD2 play a role in T cell activation, T- or NK-cell-mediated cytolysis, apoptosis in activated peripheral T cells, and regulation of T cell anergy.Host Species: Mouse
Clone: AB75
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Extracellular domain
Immunogen: Recombinant fragment encoding the external domain of the human CD2 protein
Molecular Weight: 45-58kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
Specifications
| CD2 (LFA-2) Ab-2 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Human | |
| IgG2a κ |
| AB75 | |
| Monoclonal | |
| Mouse | |
| T-Cell | |
| Recombinant fragment encoding the external domain of the human CD2 protein | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.