Learn More
Epredia™ Lab Vision™ CD2 (LFA-2) Ab-2, Mouse Monoclonal Antibody
Obtain accurate and reproducible results in immunohistochemistry experiments with Epredia™ CD2 (LFA-2) Ab-2, Mouse Monoclonal Antibody.
Marque: Epredia™ MS-1074-S
Description
CD2, which is also known as E-rosette receptor, T11 and lymphocyte-function antigen-2 (LFA-2), is expressed on T cells, thymocytes, and subset of natural killer cells. Human CD2 functions as the receptor for sheep erythrocytes, human CD58 (LFA-3), and CD15s (Sialyl Lewis X). p56lck, p59fyn, CD3η and CD3ε are tyrosine-phosphorylated after CD2 stimulation. CD2 play a role in T cell activation, T- or NK-cell-mediated cytolysis, apoptosis in activated peripheral T cells, and regulation of T cell anergy.Host Species: Mouse
Clone: AB75
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Extracellular domain
Immunogen: Recombinant fragment encoding the external domain of the human CD2 protein
Molecular Weight: 45-58kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
Spécification
| CD2 (LFA-2) Ab-2 | |
| Monoclonal | |
| Unconjugated | |
| Recombinant fragment encoding the external domain of the human CD2 protein | |
| IVD | |
| Primary | |
| Concentrated |
| Immunohistochemistry (Paraffin) | |
| AB75 | |
| Mouse | |
| 1 mL | |
| T-Cell | |
| Human | |
| IgG2a κ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.