Learn More
CD117/c-Kit/SCF-Receptor Rabbit Polyclonal Antibody, Epredia™
Optimize staining of formalin-fixed, paraffin-embedded tissues with Epredia™ CD117/c-Kit/SCF-Receptor, Rabbit Polyclonal Antibody.
£168.00 - £678.65
Specifications
| Antigen | CD117/c-Kit/SCF-Receptor |
|---|---|
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12663387
|
Epredia
RB-9038-P0 |
100 μL | Concentrated | ||||||
|
12673387
|
Epredia
RB-9038-P1 |
500 μL | Concentrated | ||||||
|
12653387
|
Epredia
RB-9038-P |
1 mL | Concentrated |
£736.00 £678.65 / 1mL Save £57.35 7% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12693387
|
Epredia
RB-9038-R7 |
7 mL | Prediluted |
£354.00 £332.87 / 7mL Save £21.13 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12603397
|
Epredia
RB-9038-RQ |
12 mL | Ready-to-Use |
£381.00 £357.71 / 12mL Save £23.29 6% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Recognizes a protein of 145kDa, which is identified as CD117/p145kit. This rabbit polyclonal antibody does not interfere with the binding of SCF to c-kit. It precipitates both the unoccupied as well as the occupied form of c-kit. The binding of the stem cell factor (SCF) to the c-kit-encoded receptor tyrosine kinase (Type III) stimulates a variety of biochemical responses that culminate in cellular proliferation, migration, or survival. C-kit plays an important role in hematopoiesis, melanogenesis, and gametogenesis. This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Dog. Others not tested
Epitope: Cytoplasmic domain
Immunogen: A synthetic peptide from the cytoplasmic domain of human CD117/c-kit protein
Molecular Weight: 145 kDa
Positive Control: Stromal tumor, skin or tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| CD117/c-Kit/SCF-Receptor | |
| Polyclonal | |
| Rabbit | |
| A synthetic peptide from the cytoplasmic domain of human CD117 / c-kit protein |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.