missing translation for 'onlineSavingsMsg'
Learn More

CD117/c-Kit/SCF-Receptor Rabbit Polyclonal Antibody, Epredia™

Optimize staining of formalin-fixed, paraffin-embedded tissues with Epredia™ CD117/c-Kit/SCF-Receptor, Rabbit Polyclonal Antibody.

Brand:  Epredia RB-9038-RQ

 View more versions of this product

Product Code. 12603397

  • ONLINE EXCLUSIVE PRICE
    £381.00
    £357.71 / 12mL
    Save £23.29
    6% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recognizes a protein of 145kDa, which is identified as CD117/p145kit. This rabbit polyclonal antibody does not interfere with the binding of SCF to c-kit. It precipitates both the unoccupied as well as the occupied form of c-kit. The binding of the stem cell factor (SCF) to the c-kit-encoded receptor tyrosine kinase (Type III) stimulates a variety of biochemical responses that culminate in cellular proliferation, migration, or survival. C-kit plays an important role in hematopoiesis, melanogenesis, and gametogenesis. This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Dog. Others not tested

Epitope: Cytoplasmic domain

Immunogen: A synthetic peptide from the cytoplasmic domain of human CD117/c-kit protein

Molecular Weight: 145 kDa

Positive Control: Stromal tumor, skin or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD117/c-Kit/SCF-Receptor
Polyclonal
Unconjugated
A synthetic peptide from the cytoplasmic domain of human CD117 / c-kit protein
IVD
Ready-to-Use
Immunohistochemistry (Paraffin)
Polyclonal
Rabbit
12 mL
Primary
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.