missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ β-Catenin Rabbit Monoclonal Antibody

Ensure accurate and reproducible results in immunohistochemistry applications with this antibody, part of a comprehensive antibody portfolio.

Brand:  Epredia™ RM-2101-S

 View more versions of this product

Product Code. 12683597

  • ONLINE EXCLUSIVE PRICE
    £953.00
    £878.79 / 1mL
    Save £74.21
    7% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The catenins (α, β and γ) are ubiquitously expressed cytoplasmic proteins associated with E-cadherin at cellular junctions. β-catenin also binds to N-cadherin and co-immunoprecipitates with APC. Cadherin/catenin complexes are linked to the cytoskeleton via a direct association between α-actinin and α-catenin. Increased tyrosine phosphorylation can disrupt catenin-cadherin complexes, influencing cellular adhesion. Mutations in the catenin gene, CTNNB1, have been implicated in its accumulation and various forms of carcinomas.

Host Species: Rabbit

Clone: E247

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide derived from near the N-terminus of human β-catenin

Molecular Weight: 92 kDa

Positive Control: Breast CA

Cellular Localization: Cell Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Catenin, beta
Monoclonal
Unconjugated
A synthetic peptide derived from near the N-terminus of human beta-catenin
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
E247
Rabbit
1 mL
Cancer and Tumor Biology
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado